5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester 95%
Lieferant: Apollo Scientific
OR925600-1G
OR925600-250MG
APOSOR925600-1GEA
719
EUR
APOSOR925600-1G
APOSOR925600-250MG
5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester 95%
5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester
Formel:
C₇H₈ClNO₃ Molecular Weight: 189.596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24843138 CAS-Nummer: 80196-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...