2-Chloroquinolin-4-amine 95%
Lieferant: Apollo Scientific
OR925609-1G
APOSOR925609-1GEA
68.3
EUR
APOSOR925609-1G
2-Chloroquinolin-4-amine 95%
2-Chloroquinolin-4-amine
Formel:
C₉H₇ClN₂ Molecular Weight: 178,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08705643 CAS-Nummer: 80947-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...