(4-Propoxyphenyl)acetic acid 98%

Lieferant: Apollo Scientific

OR925627-1G OR925627-5G
APOSOR925627-1GEA 151 EUR
APOSOR925627-1G APOSOR925627-5G
(4-Propoxyphenyl)acetic acid 98%
(4-Propoxyphenyl)acetic acid
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00460580
CAS-Nummer: 26118-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR