5,6,7,8-Tetrahydronaphthalene-1-carbaldehyde 95%

Lieferant: Apollo Scientific

OR925656-1G
APOSOR925656-1GEA 184 EUR
APOSOR925656-1G
5,6,7,8-Tetrahydronaphthalene-1-carbaldehyde 95%
5,6,7,8-Tetrahydronaphthalene-1-carbaldehyde
Formel: C₁₁H₁₂O
Molecular Weight: 160,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15474839
CAS-Nummer: 41828-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR