5,6,7,8-Tetrahydro-2H-cinnolin-3-one 95%

Lieferant: Apollo Scientific

Synonyme: 5,6,7,8-Tetrahydrocinnolin-3-ol

OR925712-1G
APOSOR925712-1GEA 145 EUR
APOSOR925712-1G
5,6,7,8-Tetrahydro-2H-cinnolin-3-one 95%
5,6,7,8-Tetrahydro-2H-cinnolin-3-one
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00828986
CAS-Nummer: 5468-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR