4-Chlorbutylacetat 98%
Lieferant: Apollo Scientific
Synonyme:
Essigsäure(4-chlorbutyl)ester
OR925717-500G
APOSOR925717-500GEA
156
EUR
APOSOR925717-500G
4-Chlorbutylacetat 98%
4-Chlorbutylacetat
Formel:
C₆H₁₁ClO₂ Molecular Weight: 150.6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001013 CAS-Nummer: 6962-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...