3-(4-Methoxyphenyl)isoxazol-5-amine 95%

Lieferant: Apollo Scientific

OR925727-1G
APOSOR925727-1GEA 208 EUR
APOSOR925727-1G
3-(4-Methoxyphenyl)isoxazol-5-amine 95%
3-(4-Methoxyphenyl)isoxazol-5-amine
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407397
CAS-Nummer: 86685-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR