5-Methoxyisoquinoline hydrochloride 95%

Lieferant: Apollo Scientific

OR925755-5G
APOSOR925755-5GEA 162 EUR
APOSOR925755-5G
5-Methoxyisoquinoline hydrochloride 95%
5-Methoxyisoquinoline hydrochloride
Formel: C₁₀H₉NO·ClH
Molecular Weight: 195.646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22683320
CAS-Nummer: 1418117-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR