6-Chloro-n,n-dimethyl-3-pyridinamine 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-N,N-dimethylpyridin-3-amine

OR925779-1G OR925779-250MG
APOSOR925779-1GEA 328 EUR
APOSOR925779-1G APOSOR925779-250MG
6-Chloro-n,n-dimethyl-3-pyridinamine 98%
6-Chloro-n,n-dimethyl-3-pyridinamine
Formel: C₇H₉ClN₂
Molecular Weight: 156.613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234337
CAS-Nummer: 41288-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR