tert-Butyl-2-oxoazetidine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR925839-1G OR925839-250MG
APOSOR925839-1GEA 502 EUR
APOSOR925839-1G APOSOR925839-250MG
tert-Butyl-2-oxoazetidine-1-carboxylate 95%
tert-Butyl-2-oxoazetidine-1-carboxylate
Formel: C₈H₁₃NO₃
Lagertemperatur: Kühlschrank
MDL: MFCD08751345
CAS-Nummer: 1140510-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR