3-Methyl-1H-pyrazole-5-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 5-Methyl-1H-pyrazole-3-carbaldehyde

OR925890-1G
APOSOR925890-1GEA 78.3 EUR
APOSOR925890-1G
3-Methyl-1H-pyrazole-5-carbaldehyde 95%
3-Methyl-1H-pyrazole-5-carbaldehyde
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159629
CAS-Nummer: 3273-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR