L-Leucine ethyl ester hydrochloride 97%

Lieferant: Apollo Scientific

OR925903-100G
APOSOR925903-100GEA 0 EUR
APOSOR925903-100G
L-Leucine ethyl ester hydrochloride 97%
L-Leucine ethyl ester hydrochloride
Formel: C₈H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00034879
CAS-Nummer: 2743-40-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR