3-Cyan-6-methylchromon 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methylchromone-3-carbonitrile, 6-Methyl-4-oxo-4H-chromene-3-carbonitrile

OR925911-5G
APOSOR925911-5GEA 89.9 EUR
APOSOR925911-5G
3-Cyan-6-methylchromon 95%
3-Cyan-6-methylchromon
Formel: C₁₁H₇NO₂
Molecular Weight: 185.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191962
CAS-Nummer: 50743-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR