(5-Ethyl-2-methoxyphenyl)acetic acid 95%
Lieferant: Apollo Scientific
OR925914-1G
APOSOR925914-1GEA
348
EUR
APOSOR925914-1G
(5-Ethyl-2-methoxyphenyl)acetic acid 95%
(5-Ethyl-2-methoxyphenyl)acetic acid
Formel:
C₁₁H₁₄O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664724 CAS-Nummer: 383134-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...