4-Methoxy-2-(pyridin-3-yl)butanoic acid 95%

Lieferant: Apollo Scientific

OR925919-1G
APOSOR925919-1GEA 609 EUR
APOSOR925919-1G
4-Methoxy-2-(pyridin-3-yl)butanoic acid 95%
4-Methoxy-2-(pyridin-3-yl)butanoic acid
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658466
CAS-Nummer: 885277-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR