3-(Methylsulfanyl)propyl-acetat 95%

Lieferant: Apollo Scientific

Synonyme: Acetic Acid 3-(Methylthio)propyl Ester

OR925951-25G
APOSOR925951-25GEA 59.6 EUR
APOSOR925951-25G
3-(Methylsulfanyl)propyl-acetat 95%
3-(Methylthio)propyl acetate
Formel: C₆H₁₂O₂S
Molecular Weight: 148.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053113
CAS-Nummer: 16630-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR