N-Boc-endo-3-aminotropane 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl endo-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate, tert-Butyl (1S,5R)-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate

OR926114-5G
APOSOR926114-5GEA 115 EUR
APOSOR926114-5G
N-Boc-endo-3-aminotropane 97%
N-Boc-endo-3-aminotropane
Formel: C₁₂H₂₂N₂O₂
Molecular Weight: 226.315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18428077
CAS-Nummer: 207405-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR