1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
7-Amino-N-methyl-1,2,3,4-tetrahydroquinoline hydrochloride
OR926128-1G
APOSOR926128-1GEA
487
EUR
APOSOR926128-1G
1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride 95%
1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride
Formel:
C₁₀H₁₅ClN₂ Molecular Weight: 198,69 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839668 CAS-Nummer: 927684-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...