2,6-Dichloro-3H-pyrimidin-4-one 96%

Lieferant: Apollo Scientific

OR926129-1G
APOSOR926129-1GEA 159 EUR
APOSOR926129-1G
2,6-Dichloro-3H-pyrimidin-4-one 96%
2,6-Dichloro-3H-pyrimidin-4-one
Formel: C₄H₂Cl₂N₂O
Molecular Weight: 164,9775 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00955562
CAS-Nummer: 120977-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR