5-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR926154-250MG
APOSOR926154-250MGEA
142
EUR
APOSOR926154-250MG
5-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 97+%
5-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₈H₅BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11043994 CAS-Nummer: 1026201-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...