2-Chloro-5-methylthiazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR926189-1G
APOSOR926189-1GEA 68.3 EUR
APOSOR926189-1G
2-Chloro-5-methylthiazole-4-carboxylic acid 95%
2-Chloro-5-methylthiazole-4-carboxylic acid
Formel: C₅H₄ClNO₂S
Molecular Weight: 177.609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12404922
CAS-Nummer: 1194374-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR