4-Butylmorpholin 95%

Lieferant: Apollo Scientific

OR926210-25G OR926210-5G
APOSOR926210-25GEA 451 EUR
APOSOR926210-25G APOSOR926210-5G
4-Butylmorpholin 95%
4-Butylmorpholin
Formel: C₈H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00023401
CAS-Nummer: 1005-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR