3-Chloroquinoline 98%

Lieferant: Apollo Scientific

OR926222-5G
APOSOR926222-5GEA 183 EUR
APOSOR926222-5G
3-Chloroquinoline 98%
3-Chloroquinoline
Formel: C9H6ClN
Molecular Weight: 163.6036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234489
CAS-Nummer: 612-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR