3-Acetyl-2,4-dimethylpyrrole 95%
Lieferant: Apollo Scientific
OR926294-5G
APOSOR926294-5GEA
149
EUR
APOSOR926294-5G
3-Acetyl-2,4-dimethylpyrrole 95%
3-Acetyl-2,4-dimethylpyrrole
Formel:
C₈H₁₁NO Molecular Weight: 137.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005221 CAS-Nummer: 2386-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...