Ethyl-2,6-dimethylimidazo[1,2-a]pyridin-3-carboxylat

Lieferant: Apollo Scientific

OR9263-1G
APOSOR9263-1GEA 220 EUR
APOSOR9263-1G
Ethyl-2,6-dimethylimidazo[1,2-a]pyridin-3-carboxylat
Ethyl-2,6-dimethylimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₂H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 81438-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR