5-Ethoxypyridin-2-amine 95%
Lieferant: Apollo Scientific
OR926327-1G
APOSOR926327-1GEA
273
EUR
APOSOR926327-1G
5-Ethoxypyridin-2-amine 95%
5-Ethoxypyridin-2-amine
Formel:
C7H10N2O Molecular Weight: 138,1671 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11135752 CAS-Nummer: 89943-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...