2,7-Dihydroxynaphthalin 98%
Lieferant: Apollo Scientific
Synonyme:
2,7-Naphthalindiol
OR926339-1KG
APOSOR926339-1KGEA
0
EUR
APOSOR926339-1KG
2,7-Dihydroxynaphthalin 98%
2,7-Dihydroxynaphthalin
Formel:
C₁₀H₈O₂ Molecular Weight: 160.17 g/mol Siedepunkt: 375 Schmelzpunkt: 184…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004085 CAS-Nummer: 582-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...