tert-Butyl (cyanomethyl)carbamate 97%

Lieferant: Apollo Scientific

Synonyme: N-(tert-Butoxycarbonyl)-2-aminoacetonitrile

OR926356-100G
APOSOR926356-100GEA 140 EUR
APOSOR926356-100G
tert-Butyl (cyanomethyl)carbamate 97%
tert-Butyl (cyanomethyl)carbamate
Formel: C₇H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00239390
CAS-Nummer: 85363-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR