3-Methyl-2-phenylpyridin 95%

Lieferant: Apollo Scientific

Synonyme: 2-Phenyl-beta-picoline

OR926358-5G
APOSOR926358-5GEA 85.7 EUR
APOSOR926358-5G
3-Methyl-2-phenylpyridin 95%
3-Methyl-2-phenylpyridine
Formel: C₁₂H₁₁N
Molecular Weight: 169.23 g/mol
Siedepunkt: 148
Lagertemperatur: Raumtemperatur
MDL: MFCD00006283
CAS-Nummer: 10273-90-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR