5-Methylthiazole-4-carboxylic acid ethyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-methylthiazole-4-carboxylate

OR926418-1G OR926418-250MG
APOSOR926418-1GEA 78.3 EUR
APOSOR926418-1G APOSOR926418-250MG
5-Methylthiazole-4-carboxylic acid ethyl ester 97%
5-Methylthiazole-4-carboxylic acid ethyl ester
Formel: C₇H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD18427704
CAS-Nummer: 61323-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR