1-Propylpiperidin-3-amine 95%

Lieferant: Apollo Scientific

OR926443-100MG
APOSOR926443-100MGEA 248 EUR
APOSOR926443-100MG
1-Propylpiperidin-3-amine 95%
1-Propylpiperidin-3-amine
Formel: C₈H₁₈N₂
Molecular Weight: 142,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059954
CAS-Nummer: 51388-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR