Boc-3-bromo-D-phenylalanine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-3-bromo-D-phenylalanine, (R)-3-(3-Bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

OR926518-5G
APOSOR926518-5GEA 162 EUR
APOSOR926518-5G
Boc-3-bromo-D-phenylalanine 97%
Boc-3-bromo-D-phenylalanine
Formel: C₁₄H₁₈BrNO₄
Molecular Weight: 344.205 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01317704
CAS-Nummer: 261360-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR