2-Propylmalonic acid 95%

Lieferant: Apollo Scientific

OR926537-10G
APOSOR926537-10GEA 156 EUR
APOSOR926537-10G
2-Propylmalonic acid 95%
2-Propylmalonic acid
Formel: C₆H₁₀O₄
Molecular Weight: 146,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840631
CAS-Nummer: 616-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR