tert-Butyl N-ethyl-N-(2-hydroxyethyl)carbamate 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl ethyl(2-hydroxyethyl)carbamate

OR926561-25G OR926561-5G
APOSOR926561-25GEA 352 EUR
APOSOR926561-25G APOSOR926561-5G
tert-Butyl N-ethyl-N-(2-hydroxyethyl)carbamate 98%
tert-Butyl N-ethyl-N-(2-hydroxyethyl)carbamate
Formel: C₉H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04973117
CAS-Nummer: 152192-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR