2-Cyan-3-methylanilin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Cyano-3-methylaniline

OR926664-1G
APOSOR926664-1GEA 52.2 EUR
APOSOR926664-1G
2-Cyan-3-methylanilin 98%
2-Cyan-3-methylanilin
Formel: C₈H₈N₂
Molecular Weight: 132.17
Lagertemperatur: Raumtemperatur
MDL: MFCD00058686
CAS-Nummer: 56043-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR