(R)-Piperidin-3-ylmethanol hydrochloride 97%

Lieferant: Apollo Scientific

OR926711-1G OR926711-5G
APOSOR926711-1GEA 72.5 EUR
APOSOR926711-1G APOSOR926711-5G
(R)-Piperidin-3-ylmethanol hydrochloride 97%
(R)-Piperidin-3-ylmethanol hydrochloride
Formel: C₆H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD14155680
CAS-Nummer: 1124199-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR