Methyl-3-methyl-1H-pyrazole-4-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-methyl-1H-pyrazole-4-carboxylate

OR926793-1G OR926793-5G
APOSOR926793-1GEA 125 EUR
APOSOR926793-1G APOSOR926793-5G
Methyl-3-methyl-1H-pyrazole-4-carboxylate 95%
Methyl-3-methyl-1H-pyrazole-4-carboxylate
Formel: C₆H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07357354
CAS-Nummer: 23170-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR