Pyridine-4-sulphonyl chloride hydrochloride 98%

Lieferant: Apollo Scientific
Gefahr

OR926874-100MG OR926874-250MG OR926874-1G OR926874-500MG OR926874-2.5G OR926874-5G
APOSOR926874-100MGEA 329 EUR
APOSOR926874-100MG APOSOR926874-250MG APOSOR926874-1G APOSOR926874-500MG APOSOR926874-2.5G APOSOR926874-5G
Pyridine-4-sulphonyl chloride hydrochloride 98%
Pyridine-4-sulphonyl chloride hydrochloride
Formel: C₅H₄ClNO₂S·ClH
Molecular Weight: 214.07 g/mol
Schmelzpunkt: 90…100 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD04973456
CAS-Nummer: 489430-50-4
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR