6,7-Dihydro-5h-cyclopenta[c]pyridazin-3-ol 95%

Lieferant: Apollo Scientific

Synonyme: 6,7-Dihydro-2H-cyclopenta[c]pyridazin-3(5H)-one

OR926886-1G
APOSOR926886-1GEA 39.3 EUR
APOSOR926886-1G
6,7-Dihydro-5h-cyclopenta[c]pyridazin-3-ol 95%
6,7-Dihydro-5h-cyclopenta[c]pyridazin-3-ol
Formel: C₇H₈N₂O
Molecular Weight: 136,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20502855
CAS-Nummer: 122001-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR