7-Chloro-2H-1,4-benzoxazin-3(4h)-one 98%

Lieferant: Apollo Scientific

Synonyme: 7-Chloro-2H-benzo[b][1,4]oxazin-3(4H)-one

OR926962-1G
APOSOR926962-1GEA 63.8 EUR
APOSOR926962-1G
7-Chloro-2H-1,4-benzoxazin-3(4h)-one 98%
7-Chloro-2H-1,4-benzoxazin-3(4h)-one
Formel: C₈H₆ClNO₂
Molecular Weight: 183,592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03019820
CAS-Nummer: 27320-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR