1,2-Dihydroxynaphthalin 95%

Lieferant: Apollo Scientific

OR927003-1G
APOSOR927003-1GEA 171 EUR
APOSOR927003-1G
1,2-Dihydroxynaphthalin 95%
1,2-Dihydroxynaphthalin
Formel: C₁₀H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00003959
CAS-Nummer: 574-00-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR