Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate 98+%

Lieferant: Apollo Scientific
Achtung

OR927013-1G
APOSOR927013-1GEA 132 EUR
APOSOR927013-1G
Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate 98+%
Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate
Formel: C₉H₈ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11975600
CAS-Nummer: 1150566-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR