2-(1H-Imidazol-1-yl)aniline 95%

Lieferant: Apollo Scientific

OR927017-1G
APOSOR927017-1GEA 81.2 EUR
APOSOR927017-1G
2-(1H-Imidazol-1-yl)aniline 95%
2-(1H-Imidazol-1-yl)aniline
Formel: C₉H₉N₃
Molecular Weight: 159,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03274531
CAS-Nummer: 26286-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR