5-Methyl-1H-pyrrole-3-carboxylic acid 97%

Lieferant: Apollo Scientific

OR927108-1G OR927108-250MG
APOSOR927108-1GEA 217 EUR
APOSOR927108-1G APOSOR927108-250MG
5-Methyl-1H-pyrrole-3-carboxylic acid 97%
5-Methyl-1H-pyrrole-3-carboxylic acid
Formel: C₆H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09991919
CAS-Nummer: 100047-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR