tert-Butyl cyclopropanecarboxylate 95%
Lieferant: Apollo Scientific
OR927117-25G
APOSOR927117-25GEA
120
EUR
APOSOR927117-25G
tert-Butyl cyclopropanecarboxylate 95%
tert-Butyl cyclopropanecarboxylate
Formel:
C₈H₁₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836696 CAS-Nummer: 87661-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...