4-[(Methylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

Synonyme: 4-((Methylamino)methyl)phenol

OR927144-1G
APOSOR927144-1GEA 79.9 EUR
APOSOR927144-1G
4-[(Methylamino)methyl]phenol 95%
4-[(Methylamino)methyl]phenol
Formel: C₈H₁₁NO
Molecular Weight: 137.179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09046544
CAS-Nummer: 78507-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR