5-(p-Tolyl)isoxazole-3-carboxaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 5-(4-Methylphenyl)isoxazole-3-carboxaldehyde

OR927181-1G
APOSOR927181-1GEA 290 EUR
APOSOR927181-1G
5-(p-Tolyl)isoxazole-3-carboxaldehyde 95%
5-(p-Tolyl)isoxazole-3-carboxaldehyde
Formel: C₁₁H₉NO₂
Molecular Weight: 187,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06199344
CAS-Nummer: 640292-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR