N-(1-Hydroxy-2-butyl)furan-2-carboxamide 95%

Lieferant: Apollo Scientific

OR927185-250MG
APOSOR927185-250MGEA 306 EUR
APOSOR927185-250MG
N-(1-Hydroxy-2-butyl)furan-2-carboxamide 95%
N-(1-Hydroxy-2-butyl)furan-2-carboxamide
Formel: C₉H₁₃NO₃
Molecular Weight: 183.204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05883458
CAS-Nummer: 791832-35-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR