1-(Ethylamino)-3-(dimethylamino)propane 95%

Lieferant: Apollo Scientific

OR927198-1G
APOSOR927198-1GEA 364 EUR
APOSOR927198-1G
1-(Ethylamino)-3-(dimethylamino)propane 95%
1-(Ethylamino)-3-(dimethylamino)propane
Formel: C₇H₁₈N₂
Molecular Weight: 130,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051552
CAS-Nummer: 19475-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR