6-Methoxy-2-pyrazinecarboxylic acid 95%
Lieferant: Apollo Scientific
OR927218-1G
APOSOR927218-1GEA
63.8
EUR
APOSOR927218-1G
6-Methoxy-2-pyrazinecarboxylic acid 95%
6-Methoxy-2-pyrazinecarboxylic acid
Formel:
C₆H₆N₂O₃ Molecular Weight: 154,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233277 CAS-Nummer: 24005-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...